siRNA Details
| Gene Symbol: CHCHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320327.1 | NP_001307256.1 | 4 | 766 | ||
| NM_016139.3 | NP_057223.1 | 4 | 762 | |||
| GenBank mRNA | AF078845.1 | - | 4 | 626 | ||
| BC003079.2 | - | 4 | 582 | 2988156 | ||
| BC015639.2 | - | 4 | 566 | 4637353 | ||
| BC066331.1 | - | 4 | 585 | 4792941 | ||
| BC071985.1 | - | 4 | 582 | 6426679 | ||
| BC100275.1 | - | 4 | 578 | 6192342 | ||

CHCHD2
Genomic Map