siRNA Details
| Gene Symbol: COMMD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308080.1 | NP_001295009.1 | 7 | 963 | ||
| NM_016144.3 | NP_057228.1 | 7 | 936 | |||
| XM_017009553.1 | XP_016865042.1 | 7 | 1058 | |||
| GenBank mRNA | AK002147.1 | - | 7 | 912 | ||
| BC005179.1 | - | 7 | 882 | 3683093 | ||

COMMD10
Genomic Map