siRNA Details
| Gene Symbol: Stat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205313.1 | NP_001192242.1 | 25 | 4064 | ||
| NM_001205314.1 | NP_001192243.1 | 25 | 4082 | |||
| NM_009283.4 | NP_033309.3 | 25 | 4097 | |||
| GenBank mRNA | AK082706.1 | - | 25 | 4079 | ||
| AK084855.1 | - | 25 | 4031 | |||
| AK159887.1 | - | 25 | 4081 | |||
| BC042551.1 | - | 8 | 2229 | |||

Stat1
Genomic Map