siRNA Details
Gene Symbol: Zbtb20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285805.1 | NP_001272734.1 | 9 | 714 | ||
NM_019778.2 | NP_062752.2 | 7 | 661 | |||
NM_181058.1 | NP_851401.1 | 5 | 439 | |||
GenBank mRNA | AB221632.1 | - | 1 | 126 | ||
AF185576.1 | - | 5 | 484 | |||
AF194030.1 | - | 3 | 276 | |||
AK005028.2 | - | 5 | 440 | |||
AK052686.1 | - | 6 | 988 | |||
AK078664.1 | - | 423 | ||||
AY028963.1 | - | 2 | 243 | |||
BC031114.1 | - | 6 | 547 | 4971586 | ||
BC056446.1 | - | 9 | 686 | 5703319 | ||
BI659024.1 | - | 10 | 702 |
