siRNA Details
| Gene Symbol: CYB5R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302826.1 | NP_001289755.1 | 7 | 1133 | ||
| NM_001302827.1 | NP_001289756.1 | 6 | 607 | |||
| NM_016229.4 | NP_057313.2 | 7 | 787 | |||
| NR_126508.1 | 7 | 787 | ||||
| XM_005252975.4 | XP_005253032.1 | 7 | 739 | |||
| XM_006718251.2 | XP_006718314.1 | 7 | 764 | |||
| XM_011520184.2 | XP_011518486.1 | 7 | 954 | |||
| XM_011520185.2 | XP_011518487.1 | 9 | 1053 | |||
| XM_017017920.1 | XP_016873409.1 | 9 | 1047 | |||
| XM_017017921.1 | XP_016873410.1 | 9 | 1049 | |||
| XM_017017922.1 | XP_016873411.1 | 7 | 681 | |||
| GenBank mRNA | AB209000.1 | - | 3 | 4823 | ||
| AF169802.1 | - | 7 | 690 | |||
| AK290102.1 | - | 7 | 1106 | |||
| AL133582.1 | - | 6 | 523 | |||
| AY665398.1 | - | 6 | 541 | |||
| BC001346.1 | - | 7 | 661 | 3451742 | ||
| BQ431043.1 | - | 6 | 545 | |||

CYB5R2
Genomic Map