siRNA Details
| Gene Symbol: GPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164372.1 | NP_001157844.1 | 3 | 387 | ||
| NM_001164373.1 | NP_001157845.1 | 3 | 296 | |||
| NM_016301.3 | NP_057385.3 | 3 | 266 | |||
| XM_005253896.4 | XP_005253953.3 | 4 | 721 | |||
| XM_017019394.1 | XP_016874883.1 | 3 | 201 | |||
| GenBank mRNA | AF117229.1 | - | 3 | 216 | ||
| AK223209.1 | - | 3 | 238 | |||
| AK314946.1 | - | 3 | 239 | |||
| AM992619.1 | - | 3 | 185 | |||
| AM992620.1 | - | 3 | 185 | |||
| AY359078.1 | - | 3 | 267 | |||
| AY364262.1 | - | 3 | 185 | |||
| BC008416.1 | - | 3 | 241 | 4048678 | ||
| BC031024.1 | - | 3 | 242 | 4695834 | ||
| BI546381.1 | - | 3 | 390 | |||
| BX537973.1 | - | 3 | 277 | |||
| DB455987.1 | - | 3 | 266 | |||

GPN3
Genomic Map