siRNA Details
Gene Symbol: TCEAL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006612.1 | NP_001006613.1 | 3 | 387 | ||
NM_001006613.1 | NP_001006614.1 | 2 | 311 | |||
NM_001006614.1 | NP_001006615.1 | 2 | 308 | |||
NM_016303.2 | NP_057387.1 | 3 | 390 | |||
GenBank mRNA | AF125535.1 | - | 3 | 348 | ||
AK312190.1 | - | 3 | 336 | |||
AL833152.1 | - | 1 | 1254 | |||
BC023544.2 | - | 3 | 341 | 4122244 | ||
BG400091.1 | - | 3 | 391 |
