siRNA Details
Gene Symbol: OR5T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004747.1 | NP_001004747.1 | 1 | 373 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
