siRNA Details
| Gene Symbol: ZNF81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007137.3 | NP_009068.2 | 3 | 337 | ||
| XM_005272600.3 | XP_005272657.1 | 3 | 395 | |||
| XM_011543899.2 | XP_011542201.1 | 3 | 2001 | |||
| XM_011543900.2 | XP_011542202.1 | 4 | 456 | |||
| XM_017029486.1 | XP_016884975.1 | 4 | 514 | |||
| XM_017029487.1 | XP_016884976.1 | 3 | 747 | |||
| GenBank mRNA | AK126949.1 | - | 4 | 456 | ||
| AY487248.1 | - | 2 | 88 | |||
| BC039609.1 | - | 4 | 432 | |||
| DA219953.1 | - | 4 | 456 | |||
| DR002896.1 | - | 3 | 325 | |||

ZNF81
Genomic Map