siRNA Details
| Gene Symbol: GSE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134473.2 | NP_001127945.1 | 15 | 4139 | ||
| NM_001278184.1 | NP_001265113.1 | 15 | 4193 | |||
| NM_014615.3 | NP_055430.1 | 16 | 4412 | |||
| XM_005255859.4 | XP_005255916.3 | 17 | 6554 | |||
| XM_005255860.3 | XP_005255917.3 | 16 | 6447 | |||
| XM_005255861.4 | XP_005255918.3 | 16 | 6335 | |||
| XM_005255863.4 | XP_005255920.1 | 16 | 4304 | |||
| XM_005255864.4 | XP_005255921.1 | 16 | 5036 | |||
| XM_005255865.4 | XP_005255922.1 | 15 | 4817 | |||
| XM_011522965.2 | XP_011521267.1 | 16 | 4803 | |||
| XM_011522966.2 | XP_011521268.1 | 16 | 4270 | |||
| XM_017023083.1 | XP_016878572.1 | 16 | 4562 | |||
| GenBank mRNA | AK127344.1 | - | 5 | 2816 | ||
| BC000753.2 | - | 4 | 1381 | |||
| BC009854.2 | - | 3 | 879 | |||
| BC037556.1 | - | 15 | 4162 | 4994198 | ||
| D80004.2 | - | 15 | 4077 | |||

GSE1
Genomic Map