siRNA Details
| Gene Symbol: RGS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286692.1 | NP_001273621.1 | 22 | 3493 | ||
| NM_001286693.1 | NP_001273622.1 | 20 | 3388 | |||
| NM_015668.4 | NP_056483.3 | 22 | 3529 | |||
| XM_005250856.2 | XP_005250913.1 | 21 | 3216 | |||
| XM_005250857.2 | XP_005250914.1 | 22 | 3375 | |||
| XM_005250858.2 | XP_005250915.1 | 21 | 3213 | |||
| XM_006716540.2 | XP_006716603.1 | 22 | 3643 | |||
| XM_011516957.1 | XP_011515259.1 | 23 | 3824 | |||
| XM_011516958.1 | XP_011515260.1 | 22 | 3722 | |||
| XM_017013309.1 | XP_016868798.1 | 21 | 3216 | |||
| XM_017013310.1 | XP_016868799.1 | 20 | 3054 | |||
| XM_017013311.1 | XP_016868800.1 | 15 | 2072 | |||
| GenBank mRNA | AK097399.1 | - | 11 | 1562 | ||
| AK292559.1 | - | 22 | 3339 | |||
| AL117544.1 | - | 3 | 354 | |||
| AY009106.1 | - | 22 | 3322 | |||
| BC036665.1 | - | 22 | 3380 | 5268331 | ||
| BC092411.1 | - | 22 | 3526 | 30386231 | ||

RGS22
Genomic Map