siRNA Details
| Gene Symbol: ATPIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016311.4 | NP_057395.1 | 3 | 292 | ||
| NM_178190.2 | NP_835497.1 | 3 | 368 | |||
| NM_178191.2 | NP_835498.1 | 2 | 1676 | |||
| GenBank mRNA | AB029042.1 | - | 3 | 220 | ||
| AF114836.1 | - | 378 | ||||
| AK000934.1 | - | 2 | 1613 | |||
| AK316600.1 | - | 3 | 279 | |||
| AL050386.1 | - | 2 | 1606 | |||
| AL583540.3 | - | 3 | 291 | |||
| BC004955.1 | - | 3 | 175 | 3625379 | ||
| BC009677.1 | - | 3 | 250 | 3877506 | ||
| BG036978.1 | - | 3 | 340 | |||
| BG337531.1 | - | 3 | 248 | |||
| BP271519.1 | - | 3 | 288 | |||
| BT009849.1 | - | 3 | 199 | |||
| CR457097.1 | - | 3 | 199 | |||

ATPIF1
Genomic Map