siRNA Details
| Gene Symbol: HSD17B14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016246.2 | NP_057330.2 | 6 | 674 | ||
| XM_005258969.3 | XP_005259026.1 | 6 | 683 | |||
| XM_005258970.3 | XP_005259027.1 | 5 | 593 | |||
| GenBank mRNA | AF126781.1 | - | 6 | 520 | ||
| AK307437.1 | - | 6 | 483 | |||
| AY358430.1 | - | 6 | 513 | |||
| BC006283.2 | - | 6 | 467 | 4026397 | ||
| BC006294.1 | - | 6 | 475 | 4025809 | ||

HSD17B14
Genomic Map