siRNA Details
| Gene Symbol: RBMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002897.4 | NP_002888.1 | 14 | 1765 | ||
| NM_016836.3 | NP_058520.1 | 14 | 1774 | |||
| XM_005246737.1 | XP_005246794.1 | 15 | 1533 | |||
| XM_005246738.2 | XP_005246795.1 | 15 | 1868 | |||
| XM_005246739.2 | XP_005246796.1 | 15 | 1738 | |||
| XM_005246740.1 | XP_005246797.1 | 15 | 1524 | |||
| XM_005246741.3 | XP_005246798.1 | 15 | 1735 | |||
| XM_017004626.1 | XP_016860115.1 | 14 | 1811 | |||
| GenBank mRNA | AK127157.1 | - | 5 | 2166 | ||
| AK297812.1 | - | 15 | 1524 | |||
| BC012992.2 | - | 14 | 1460 | 3530897 | ||
| BC012993.2 | - | 14 | 1460 | 3530898 | ||
| BC018951.2 | - | 14 | 1761 | 4137722 | ||
| BC051889.1 | - | 14 | 1347 | |||
| BX648246.1 | - | 7 | 8931 | |||
| L11289.1 | - | 14 | 1397 | |||
| X64652.1 | - | 13 | 1280 | |||
| X77494.1 | - | 16 | 1510 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001141931, NM_020296, NM_001141932, AK135280, BC016501, and others.

RBMS1
Genomic Map