siRNA Details
| Gene Symbol: ANKRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286721.1 | NP_001273650.1 | 5 | 935 | ||
| NM_017664.3 | NP_060134.2 | 4 | 841 | |||
| NR_104586.1 | 5 | 973 | ||||
| NR_104587.1 | 5 | 935 | ||||
| GenBank mRNA | AK000100.1 | - | 4 | 803 | ||
| AK094222.1 | - | 1 | 841 | |||
| BC001727.1 | - | 5 | 834 | 3354202 | ||
| BC039715.1 | - | 5 | 903 | |||
| BX354283.2 | - | 5 | 832 | |||

ANKRD10
Genomic Map