siRNA Details
| Gene Symbol: FAM8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016255.2 | NP_057339.1 | 1, 2 | 843 | ||
| GenBank mRNA | AF097027.1 | - | 1, 2 | 775 | ||
| AK296393.1 | - | 2, 3 | 157 | |||
| AK312814.1 | - | 1, 2 | 778 | |||
| BC047881.1 | - | 1, 2 | 767 | 5742372 | ||

FAM8A1
Genomic Map