siRNA Details
| Gene Symbol: C7orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303470.1 | NP_001290399.1 | 9 | 1046 | ||
| NM_018275.4 | NP_060745.3 | 9 | 1546 | |||
| GenBank mRNA | AK001787.1 | - | 5 | 620 | ||
| AK054872.1 | - | 9 | 1042 | |||
| AK302342.1 | - | 7 | 894 | |||
| AL136555.1 | - | 2 | 245 | |||
| BC002749.2 | - | 9 | 946 | |||
| BC015722.2 | - | 9 | 1504 | 3920412 | ||

C7orf43
Genomic Map