siRNA Details
| Gene Symbol: HPCAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282396.1 | NP_001269325.1 | 4, 5 | 680 | ||
| NM_016257.3 | NP_057341.1 | 3, 4 | 799 | |||
| XM_017001447.1 | XP_016856936.1 | 3, 4 | 412 | |||
| GenBank mRNA | AB001105.1 | - | 3, 4 | 781 | ||
| AK294412.1 | - | 4, 5 | 495 | |||
| AK312314.1 | - | 3, 4 | 521 | |||
| AL136591.1 | - | 3, 4 | 506 | |||
| BC030827.1 | - | 3, 4 | 503 | 4811834 | ||

HPCAL4
Genomic Map