siRNA Details
| Gene Symbol: ZNF695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204221.1 | NP_001191150.1 | 2 | 315 | ||
| NM_020394.4 | NP_065127.4 | 2 | 315 | |||
| NR_037892.1 | 2 | 315 | ||||
| GenBank mRNA | BC023527.2 | - | 2 | 223 | ||
| BC041082.1 | - | 2 | 252 | |||
| BC055096.1 | - | 2 | 279 | |||
| CN394881.1 | - | 2 | 247 | |||
| DA809929.1 | - | 2 | 315 | |||

ZNF695
Genomic Map