siRNA Details
| Gene Symbol: EPB41L4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022140.3 | NP_071423.3 | 15 | 1554 | ||
| XM_011543530.1 | XP_011541832.1 | 15 | 1731 | |||
| XM_011543531.2 | XP_011541833.1 | 15 | 1731 | |||
| XM_011543532.1 | XP_011541834.1 | 15 | 1731 | |||
| XM_011543533.1 | XP_011541835.1 | 14 | 1647 | |||
| XM_017009689.1 | XP_016865178.1 | 14 | 1647 | |||
| GenBank mRNA | AB030240.1 | - | 15 | 1286 | ||
| AK096057.1 | - | 4 | 236 | |||
| BC114632.1 | - | 15 | 1380 | |||
| BC114942.1 | - | 15 | 1375 | |||
| CB048933.1 | - | 2 | 109 | |||

EPB41L4A
Genomic Map