siRNA Details
| Gene Symbol: TMEM135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168724.1 | NP_001162195.1 | 1 | 321 | ||
| NM_022918.3 | NP_075069.3 | 1 | 321 | |||
| NR_033149.1 | 1 | 321 | ||||
| XM_017018140.1 | XP_016873629.1 | 1 | 314 | |||
| GenBank mRNA | AK291804.1 | - | 1 | 231 | ||
| AK302473.1 | - | 1 | 125 | |||
| BC030952.1 | - | 1 | 180 | |||
| BC051462.1 | - | 1 | 142 | 5278478 | ||
| BX648678.1 | - | 1 | 307 | |||

TMEM135
Genomic Map