siRNA Details
| Gene Symbol: FRY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023037.2 | NP_075463.2 | 8 | 1261 | ||
| XM_006719749.3 | XP_006719812.1 | 8 | 2342 | |||
| XM_017020303.1 | XP_016875792.1 | 8 | 2819 | |||
| XM_017020304.1 | XP_016875793.1 | 8 | 1261 | |||
| XM_017020305.1 | XP_016875794.1 | 8 | 2838 | |||
| GenBank mRNA | AL049784.1 | - | 8 | 1261 | ||
| U50534.1 | - | 8 | 1260 | |||

FRY
Genomic Map