siRNA Details
| Gene Symbol: EMILIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032048.2 | NP_114437.2 | 8 | 3449 | ||
| GenBank mRNA | AB026706.1 | - | 4 | 755 | ||
| AF270513.1 | - | 8 | 3449 | |||
| AK090519.1 | - | 5 | 1544 | |||
| BC032231.1 | - | 6 | 1122 | |||
| BC136541.1 | - | 8 | 3403 | 9020531 | ||

EMILIN2
Genomic Map