siRNA Details
| Gene Symbol: QRICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032134.2 | NP_115510.1 | 8 | 3887 | ||
| NR_130649.1 | 8 | 978 | ||||
| XM_005257728.3 | XP_005257785.1 | 8 | 4897 | |||
| XM_006722136.3 | XP_006722199.1 | 4 | 1000 | |||
| XM_011525344.2 | XP_011523646.1 | 6 | 3570 | |||
| XM_017025206.1 | XP_016880695.1 | 8 | 4897 | |||
| XM_017025207.1 | XP_016880696.1 | 7 | 4837 | |||
| XM_017025208.1 | XP_016880697.1 | 6 | 968 | |||
| GenBank mRNA | AK058102.1 | - | 5 | 731 | ||
| AL136774.1 | - | 8 | 3887 | |||
| BC131559.1 | - | 8 | 863 | |||

QRICH2
Genomic Map