siRNA Details
| Gene Symbol: DQX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133637.2 | NP_598376.2 | 4 | 960 | ||
| XM_011532644.1 | XP_011530946.1 | 3 | 639 | |||
| XM_011532645.1 | XP_011530947.1 | 1 | 72 | |||
| XM_011532646.2 | XP_011530948.1 | 4 | 960 | |||
| XM_017003520.1 | XP_016859009.1 | 2 | 413 | |||
| GenBank mRNA | AK074337.1 | - | 4 | 867 | ||
| AK092420.1 | - | 4 | 961 | |||
| BC075018.2 | - | 2 | 487 | 30915408 | ||

DQX1
Genomic Map