siRNA Details
Gene Symbol: GAS2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139285.3 | NP_644814.1 | 1 | 95 | ||
GenBank mRNA | AF508784.1 | - | 1 | 95 | ||
AF508785.1 | - | 1 | 95 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GAS2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139285.3 | NP_644814.1 | 1 | 95 | ||
GenBank mRNA | AF508784.1 | - | 1 | 95 | ||
AF508785.1 | - | 1 | 95 |