siRNA Details
| Gene Symbol: GAS2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139285.3 | NP_644814.1 | 1, 2 | 416 | ||
| GenBank mRNA | AF508784.1 | - | 1, 2 | 416 | ||
| AF508785.1 | - | 1, 2 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAS2L2| Gene Symbol: GAS2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139285.3 | NP_644814.1 | 1, 2 | 416 | ||
| GenBank mRNA | AF508784.1 | - | 1, 2 | 416 | ||
| AF508785.1 | - | 1, 2 | 416 | |||