siRNA Details
| Gene Symbol: PPM1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122870.2 | NP_001116342.1 | 9 | 1307 | ||
| NM_144641.3 | NP_653242.3 | 10 | 1673 | |||
| XM_005264879.2 | XP_005264936.1 | 10 | 1766 | |||
| XM_005264880.3 | XP_005264937.1 | 9 | 1486 | |||
| XM_005264881.1 | XP_005264938.1 | 10 | 1562 | |||
| GenBank mRNA | AK056894.1 | - | 9 | 1909 | ||
| AK096681.1 | - | 8 | 1601 | |||
| AK129647.1 | - | 4 | 938 | |||
| AL832484.1 | - | 8 | 1581 | |||
| BC009644.1 | - | 9 | 1268 | 3905720 | ||

PPM1M
Genomic Map