siRNA Details
| Gene Symbol: CCDC155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144688.4 | NP_653289.3 | 12 | 1188 | ||
| XM_005258549.3 | XP_005258606.1 | 12 | 1188 | |||
| XM_011526488.2 | XP_011524790.1 | 12 | 1188 | |||
| XM_011526489.2 | XP_011524791.1 | 12 | 3088 | |||
| XM_011526490.2 | XP_011524792.1 | 12 | 1188 | |||
| XM_011526494.2 | XP_011524796.1 | 13 | 1161 | |||
| XM_011526495.2 | XP_011524797.1 | 13 | 1197 | |||
| XM_011526496.2 | XP_011524798.1 | 11 | 1049 | |||
| XM_011526497.2 | XP_011524799.1 | 12 | 1147 | |||
| XM_017026333.1 | XP_016881822.1 | 11 | 1321 | |||
| GenBank mRNA | AK057220.1 | - | 12 | 1157 | ||
| BC029811.1 | - | 12 | 1107 | 5171712 | ||

CCDC155
Genomic Map