siRNA Details
| Gene Symbol: GOT1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152413.2 | NP_689626.2 | 4 | 606 | ||
| XM_005273399.3 | XP_005273456.1 | 4 | 978 | |||
| XM_006716285.3 | XP_006716348.1 | 4 | 979 | |||
| GenBank mRNA | BC029504.1 | - | 4 | 607 | 5273324 | |
| BM559469.1 | - | 607 | ||||

GOT1L1
Genomic Map