siRNA Details
| Gene Symbol: GPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164372.1 | NP_001157844.1 | 3 | 436 | ||
| NM_001164373.1 | NP_001157845.1 | 3 | 345 | |||
| NM_016301.3 | NP_057385.3 | 3 | 315 | |||
| XM_005253896.4 | XP_005253953.3 | 4 | 770 | |||
| XM_017019394.1 | XP_016874883.1 | 3 | 250 | |||
| GenBank mRNA | AF117229.1 | - | 3 | 265 | ||
| AK223209.1 | - | 3 | 287 | |||
| AK314946.1 | - | 3 | 288 | |||
| AM992619.1 | - | 3 | 234 | |||
| AM992620.1 | - | 3 | 234 | |||
| AY359078.1 | - | 3 | 316 | |||
| AY364262.1 | - | 3 | 234 | |||
| BC008416.1 | - | 3 | 290 | 4048678 | ||
| BC031024.1 | - | 3 | 291 | 4695834 | ||
| BI546381.1 | - | 3 | 439 | |||
| BX537973.1 | - | 3 | 326 | |||

GPN3
Genomic Map