siRNA Details
| Gene Symbol: TCEAL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006612.1 | NP_001006613.1 | 3 | 417 | ||
| NM_001006613.1 | NP_001006614.1 | 2 | 341 | |||
| NM_001006614.1 | NP_001006615.1 | 2 | 338 | |||
| NM_016303.2 | NP_057387.1 | 3 | 420 | |||
| GenBank mRNA | AF125535.1 | - | 3 | 378 | ||
| AK312190.1 | - | 3 | 366 | |||
| AL833152.1 | - | 1 | 1284 | |||
| BC023544.2 | - | 3 | 371 | 4122244 | ||
| BG400091.1 | - | 3 | 421 | |||

TCEAL9
Genomic Map