siRNA Details
| Gene Symbol: ZNF596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042415.2 | NP_001035880.1 | 6 | 1080 | ||
| NM_001042416.2 | NP_001035881.1 | 6 | 1213 | |||
| NM_001287254.1 | NP_001274183.1 | 6 | 1071 | |||
| NM_001287255.1 | NP_001274184.1 | 6 | 1046 | |||
| NM_001287256.1 | NP_001274185.1 | 6 | 1042 | |||
| NM_001287399.1 | NP_001274328.1 | 6 | 1129 | |||
| NM_173539.3 | NP_775810.2 | 6 | 1131 | |||
| XM_011534744.1 | XP_011533046.1 | 6 | 1053 | |||
| XM_017013166.1 | XP_016868655.1 | 5 | 2770 | |||
| GenBank mRNA | AK313454.1 | - | 6 | 954 | ||
| BC026190.1 | - | 6 | 954 | 4837032 | ||

ZNF596
Genomic Map