siRNA Details
| Gene Symbol: C8orf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173687.3 | NP_775958.1 | 5 | 790 | ||
| NR_126398.1 | 7 | 972 | ||||
| NR_126399.1 | 5 | 781 | ||||
| GenBank mRNA | AK094450.1 | - | 5 | 738 | ||
| BC063497.1 | - | 5 | 692 | |||
| BC073830.1 | - | 7 | 920 | |||
| BC120993.2 | - | 5 | 716 | |||

C8orf31
Genomic Map