siRNA Details
| Gene Symbol: SHISA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145176.1 | NP_001138648.1 | 4 | 2453 | ||
| GenBank mRNA | AK127223.1 | - | 1 | 624 | ||
| BC093900.1 | - | 1 | 614 | |||
| BC112197.1 | - | 1 | 614 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHISA7| Gene Symbol: SHISA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145176.1 | NP_001138648.1 | 4 | 2453 | ||
| GenBank mRNA | AK127223.1 | - | 1 | 624 | ||
| BC093900.1 | - | 1 | 614 | |||
| BC112197.1 | - | 1 | 614 | |||