siRNA Details
| Gene Symbol: LRRTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178011.4 | NP_821079.3 | 3 | 3026 | ||
| NR_111909.1 | 3 | 1070 | ||||
| GenBank mRNA | AK294029.1 | - | 2 | 1246 | ||
| BC111492.1 | - | 3 | 2978 | 5196989 | ||
| BX640611.1 | - | 3 | 3004 | |||

LRRTM3
Genomic Map