siRNA Details
Gene Symbol: ZNF678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178549.3 | NP_848644.2 | 4 | 1835 | ||
NR_033184.1 | 4 | 1863 | ||||
GenBank mRNA | BC042500.1 | - | 4 | 1839 | 4823179 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178549.3 | NP_848644.2 | 4 | 1835 | ||
NR_033184.1 | 4 | 1863 | ||||
GenBank mRNA | BC042500.1 | - | 4 | 1839 | 4823179 |