siRNA Details
| Gene Symbol: ZNF678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178549.3 | NP_848644.2 | 4 | 544 | ||
| NR_033184.1 | 4 | 572 | ||||
| NR_102302.1 | 5 | 665 | ||||
| GenBank mRNA | AK307248.1 | - | 5 | 641 | ||
| AK311313.1 | - | 4 | 491 | |||
| BC042500.1 | - | 4 | 548 | 4823179 | ||
| CD655825.1 | - | 4 | 479 | |||

ZNF678
Genomic Map