siRNA Details
| Gene Symbol: LRRTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178839.4 | NP_849161.2 | 2 | 1726 | ||
| XM_017003986.1 | XP_016859475.1 | 2 | 2258 | |||
| XM_017003987.1 | XP_016859476.1 | 2 | 2106 | |||
| GenBank mRNA | AK056644.1 | - | 2 | 1757 | ||
| AK290512.1 | - | 2 | 1976 | |||
| AY182024.1 | - | 1 | 1456 | |||
| AY358310.1 | - | 2 | 2175 | |||
| BC045113.1 | - | 2 | 1726 | 5200499 | ||

LRRTM1
Genomic Map