siRNA Details
| Gene Symbol: SBSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166034.1 | NP_001159506.1 | 2 | 1721 | ||
| NM_001166035.1 | NP_001159507.1 | 2 | 458 | |||
| NM_198538.3 | NP_940940.1 | 3 | 692 | |||
| XM_011526929.2 | XP_011525231.1 | 3 | 1687 | |||
| XM_011526930.2 | XP_011525232.1 | 3 | 1643 | |||
| XM_011526931.2 | XP_011525233.1 | 3 | 1633 | |||
| GenBank mRNA | AA055367.1 | - | 2 | 244 | ||
| AK291305.1 | - | 3 | 652 | |||
| AY358701.1 | - | 3 | 692 | |||
| BC063640.1 | - | 3 | 634 | 4750640 | ||
| BX648076.1 | - | 2 | 1683 | |||
| CU448844.1 | - | 2 | 674 | |||

SBSN
Genomic Map