siRNA Details
| Gene Symbol: DRAXIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198545.3 | NP_940947.3 | 7 | 1262 | ||
| XM_011541405.2 | XP_011539707.1 | 8 | 1436 | |||
| XM_017001214.1 | XP_016856703.1 | 8 | 1572 | |||
| GenBank mRNA | AK075558.1 | - | 7 | 1257 | ||
| AK311439.1 | - | 4 | 410 | |||
| AY358750.1 | - | 7 | 1223 | |||
| BC021286.2 | - | 5 | 612 | |||
| BC111064.1 | - | 7 | 1264 | 5769984 | ||

DRAXIN
Genomic Map