siRNA Details
| Gene Symbol: SIMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308195.1 | NP_001295124.1 | 6 | 2076 | ||
| NM_001308196.1 | NP_001295125.1 | 8 | 2284 | |||
| NM_001308200.1 | NP_001295129.1 | 3 | 532 | |||
| NM_198567.5 | NP_940969.3 | 5 | 774 | |||
| NR_131772.1 | 5 | 774 | ||||
| XM_011534553.2 | XP_011532855.1 | 7 | 6081 | |||
| XM_011534554.2 | XP_011532856.1 | 7 | 1953 | |||
| XM_011534556.2 | XP_011532858.1 | 6 | 5950 | |||
| XM_017009454.1 | XP_016864943.1 | 6 | 1884 | |||
| XM_017009455.1 | XP_016864944.1 | 7 | 2194 | |||
| XM_017009456.1 | XP_016864945.1 | 7 | 2090 | |||
| GenBank mRNA | BC032390.1 | - | 3 | 500 | ||
| BC037298.1 | - | 8 | 2285 | 4823991 | ||
| BC066980.1 | - | 5 | 767 | 4821958 | ||
| BC073880.1 | - | 3 | 532 | |||
| KF151170.1 | - | 6 | 1944 | |||

SIMC1
Genomic Map