siRNA Details
| Gene Symbol: NBEAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114132.1 | NP_001107604.1 | 54 | 8211 | ||
| XM_005246787.3 | XP_005246844.1 | 54 | 8237 | |||
| XM_005246788.2 | XP_005246845.1 | 33 | 4713 | |||
| XM_006712698.3 | XP_006712761.1 | 55 | 8324 | |||
| XM_006712699.3 | XP_006712762.1 | 55 | 8324 | |||
| XM_011511658.2 | XP_011509960.1 | 55 | 8279 | |||
| XM_011511659.2 | XP_011509961.1 | 55 | 8336 | |||
| XM_011511660.2 | XP_011509962.1 | 51 | 7658 | |||
| XM_011511662.2 | XP_011509964.1 | 47 | 6993 | |||
| XM_017004728.1 | XP_016860217.1 | 42 | 6391 | |||
| XM_017004729.1 | XP_016860218.1 | 30 | 4405 | |||
| GenBank mRNA | AB053319.1 | - | 4 | 462 | ||
| AK127772.1 | - | 27 | 3809 | |||
| AK128636.1 | - | 24 | 2938 | |||
| AY172970.1 | - | 25 | 3117 | |||
| BX647133.1 | - | 18 | 2038 | |||

NBEAL1
Genomic Map