siRNA Details
| Gene Symbol: TMEFF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305134.1 | NP_001292063.1 | 9 | 1326 | ||
| NM_016192.3 | NP_057276.2 | 9 | 1326 | |||
| XM_011510890.2 | XP_011509192.1 | 8 | 1294 | |||
| XM_017003739.1 | XP_016859228.1 | 8 | 1306 | |||
| GenBank mRNA | AB004064.1 | - | 995 | |||
| AB017269.1 | - | 9 | 1284 | |||
| AF179274.2 | - | 9 | 1311 | |||
| AF242222.1 | - | 9 | 1326 | |||
| AK074507.1 | - | 9 | 940 | |||
| AK074632.1 | - | 9 | 1309 | |||
| AK222691.1 | - | 9 | 1331 | |||
| AK315376.1 | - | 9 | 1327 | |||
| AL157430.1 | - | 9 | 1309 | |||
| AY358907.1 | - | 9 | 1292 | |||
| BC008973.1 | - | 9 | 1267 | 4180709 | ||
| CR457390.1 | - | 9 | 917 | |||

TMEFF2
Genomic Map