siRNA Details
| Gene Symbol: LRRC75A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113567.2 | NP_001107039.1 | 4 | 2739 | ||
| NM_207387.3 | NP_997270.2 | 3 | 2623 | |||
| XM_011523845.2 | XP_011522147.1 | 5 | 2812 | |||
| XM_017024619.1 | XP_016880108.1 | 4 | 2508 | |||
| XM_017024620.1 | XP_016880109.1 | 5 | 2671 | |||
| GenBank mRNA | AK093015.1 | - | 3 | 2282 | ||
| AL833245.1 | - | 3 | 2649 | |||
| AL833260.1 | - | 3 | 2649 | |||

LRRC75A
Genomic Map