siRNA Details
| Gene Symbol: ZNF517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213605.2 | NP_998770.2 | 3, 4 | 272 | ||
| NR_134284.1 | 3, 4 | 272 | ||||
| XM_011517014.2 | XP_011515316.1 | 3, 4 | 811 | |||
| XM_011517015.2 | XP_011515317.1 | 3, 4 | 253 | |||
| GenBank mRNA | AK096527.1 | - | 3, 4 | 811 | ||
| AK131440.1 | - | 3, 4 | 272 | |||
| BC126161.1 | - | 2, 3 | 192 | 8991877 | ||
| DB016003.1 | - | 3, 4 | 272 | |||

ZNF517
Genomic Map