siRNA Details
| Gene Symbol: ROR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005012.3 | NP_005003.2 | 9 | 2282 | ||
| XM_011541526.1 | XP_011539828.1 | 9 | 1881 | |||
| XM_017001376.1 | XP_016856865.1 | 8 | 2182 | |||
| XM_017001377.1 | XP_016856866.1 | 11 | 4825 | |||
| GenBank mRNA | AK309189.1 | - | 6 | 1524 | ||
| BC128386.1 | - | 10 | 1892 | |||
| M97675.1 | - | 9 | 2258 | |||

ROR1
Genomic Map