siRNA Details
| Gene Symbol: VGLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320494.1 | NP_001307423.1 | 4 | 9729 | ||
| NM_016206.3 | NP_057290.2 | 4 | 9888 | |||
| XM_006713138.3 | XP_006713201.1 | 4 | 10055 | |||
| GenBank mRNA | AF099505.1 | - | 1 | 1022 | ||
| AK095826.1 | - | 1 | 2269 | |||
| BC017383.1 | - | 2 | 620 | |||
| BC062478.1 | - | 1 | 938 | |||

VGLL3
Genomic Map