siRNA Details
| Gene Symbol: ACVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105.4 | NP_001096.1 | 11 | 2443 | ||
| NM_001111067.2 | NP_001104537.1 | 11 | 2262 | |||
| XM_005246939.3 | XP_005246996.1 | 11 | 2314 | |||
| XM_005246940.3 | XP_005246997.1 | 11 | 2282 | |||
| XM_006712825.3 | XP_006712888.1 | 11 | 2395 | |||
| XM_011512106.2 | XP_011510408.1 | 12 | 2362 | |||
| XM_011512107.2 | XP_011510409.1 | 11 | 2334 | |||
| XM_011512108.2 | XP_011510410.1 | 11 | 3741 | |||
| GenBank mRNA | BC033867.1 | - | 11 | 2443 | 5209711 | |
| BC036748.1 | - | 11 | 2443 | |||
| Z22534.1 | - | 10 | 2116 | |||

ACVR1
Genomic Map