siRNA Details
| Gene Symbol: NEK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204182.1 | NP_001191111.1 | 3 | 660 | ||
| NM_001204183.1 | NP_001191112.1 | 3 | 660 | |||
| NM_002497.3 | NP_002488.1 | 3 | 660 | |||
| XM_005273147.1 | XP_005273204.1 | 3 | 657 | |||
| GenBank mRNA | BC043502.1 | - | 3 | 648 | 5541026 | |
| BC052807.1 | - | 3 | 655 | |||
| BT019729.1 | - | 3 | 509 | |||
| U11050.1 | - | 3 | 643 | |||

NEK2
Genomic Map